Learn More
Biomatik Corporation Rat Bone Morphogenetic Protein Receptor 1A (BMPR1A) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54129
The Rat Bone Morphogenetic Protein Receptor 1A (BMPR1A) ELISA Kit allows for quantitative determination in tissue homogenates and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 0.781-50 ng/mL. Sensitivity: 0.24 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q78EA7. NCBI Gene ID: 81507. Restriction: For Research Use Only.
Catalog No. 50-150-1206