Learn More
Biomatik Corporation Rat Endocrine Gland Derived Vascular Endothelial Growth Factor (EG-VEGF) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54223
The Rat Endocrine Gland Derived Vascular Endothelial Growth Factor (EG-VEGF) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 31.25-2000 pg/mL. Sensitivity: 13.6 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q8R414. NCBI Gene ID: 192205. Restriction: For Research Use Only.
Catalog No. 50-150-1225