Learn More
Biomatik Corporation Chicken Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54272
The Chicken Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Chicken Gallus. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P48800. NCBI Gene ID: 396413. Restriction: For Research Use Only.
Catalog No. 50-150-1242