Learn More
Biomatik Corporation Mouse Fibroblast Growth Factor 4 (FGF4) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54291
The Mouse Fibroblast Growth Factor 4 (FGF4) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 15.625-1000 pg/mL. Sensitivity: 5.4 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P11403. NCBI Gene ID: 14175. Restriction: For Research Use Only.
Catalog No. 50-150-1251