Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 4 (FGF4) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54292
The Rat Fibroblast Growth Factor 4 (FGF4) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 15.625-1000 pg/mL. Sensitivity: 5.4 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q8R5L6. NCBI Gene ID: 116499. Restriction: For Research Use Only.
Catalog No. 50-150-1252