Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 9 (FGF9) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54307
The Rat Fibroblast Growth Factor 9 (FGF9) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 78.125-5000 pg/mL. Sensitivity: 37 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P36364. NCBI Gene ID: 25444. Restriction: For Research Use Only.
Catalog No. 50-150-1263