Learn More
Biomatik Corporation Human Fibroblast Growth Factor Receptor 3 (FGFR3) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKL54312
The Human Fibroblast Growth Factor Receptor 3 (FGFR3) ELISA Kit allows for quantitative determination in tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P22607. NCBI Gene ID: 2261. Restriction: For Research Use Only.
Catalog No. 50-150-1267