Learn More
Biomatik Corporation Human Interleukin 1 Delta (FIL1d) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54316
The Human Interleukin 1 Delta (FIL1d) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q9UBH0. NCBI Gene ID: 26525. Restriction: For Research Use Only.
Catalog No. 50-150-1270