Learn More
Biomatik Corporation Human Glucocorticoid Induced Tumor Necrosis Factor Receptor (GITR) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54341
The Human Glucocorticoid Induced Tumor Necrosis Factor Receptor (GITR) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 5.9 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q9Y5U5. NCBI Gene ID: 8784. Restriction: For Research Use Only.
Catalog No. 50-150-1283