Learn More
Biomatik Corporation Rat Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54364
The Rat Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q06175. NCBI Gene ID: 25433. Restriction: For Research Use Only.
Catalog No. 50-150-1291