Learn More
Biomatik Corporation Mouse Hepatocyte Growth Factor (HGF) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54371
The Mouse Hepatocyte Growth Factor (HGF) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q08048. NCBI Gene ID: 15234. Restriction: For Research Use Only.
Catalog No. 50-150-1295