Learn More
Biomatik Corporation Human Hepatocyte Growth Factor Activator (HGFAC) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54374
The Human Hepatocyte Growth Factor Activator (HGFAC) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q04756. NCBI Gene ID: 3083. Restriction: For Research Use Only.
Catalog No. 50-150-1297