Learn More
Biomatik Corporation Human Hypoxia Inducible Factor 1 Alpha (HIF1a) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54376
The Human Hypoxia Inducible Factor 1 Alpha (HIF1a) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.055 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q16665. NCBI Gene ID: 3091. Restriction: For Research Use Only.
Catalog No. 50-150-1298