Learn More
Biomatik Corporation Rat Hypoxia Inducible Factor 1 Alpha (HIF1a) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54378
The Rat Hypoxia Inducible Factor 1 Alpha (HIF1a) ELISA Kit allows for quantitative determination in tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O35800. NCBI Gene ID: 29560. Restriction: For Research Use Only.
Catalog No. 50-150-1299