Learn More
Biomatik Corporation Rat Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKL54386
The Rat Intercellular Adhesion Molecule 1 (ICAM1) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.113 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q00238. NCBI Gene ID: 25464. Restriction: For Research Use Only.
Catalog No. 50-150-1303