Learn More
Biomatik Corporation Bovine Interferon Gamma (IFNg) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54411
The Bovine Interferon Gamma (IFNg) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine Cattle. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.3 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P07353. NCBI Gene ID: 281237. Restriction: For Research Use Only.
Catalog No. 50-150-1315