Learn More
Biomatik Corporation Rat Interferon Gamma (IFNg) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54419
The Rat Interferon Gamma (IFNg) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 15.625-1000 pg/mL. Sensitivity: 5.3 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P01581. NCBI Gene ID: 25712. Restriction: For Research Use Only.
Catalog No. 50-150-1319