Learn More
Biomatik Corporation Bovine Insulin Like Growth Factor 1 (IGF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKL54422
Encompass_Preferred
The Bovine Insulin Like Growth Factor 1 (IGF1) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine Cattle. Detection Range: 3.12-200 ng/mL. Sensitivity: 1.23 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: F2X2F1. NCBI Gene ID: 281239. Restriction: For Research Use Only.
Catalog No. 50-150-1320
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.