Learn More
Biomatik Corporation Human Insulin Like Growth Factor 2 Receptor (IGF2R) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54441
The Human Insulin Like Growth Factor 2 Receptor (IGF2R) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.625-40 ng/mL. Sensitivity: 0.244 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P11717. NCBI Gene ID: 3482. Restriction: For Research Use Only.
Catalog No. 50-150-1329