Learn More
Biomatik Corporation Rat Insulin Like Growth Factor Binding Protein 5 (IGFBP5) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54467
The Rat Insulin Like Growth Factor Binding Protein 5 (IGFBP5) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 15.625-1000 pg/mL. Sensitivity: 7.2 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P24594. NCBI Gene ID: 25285. Restriction: For Research Use Only.
Catalog No. 50-150-1343