Learn More
Biomatik Corporation Mouse Insulin Like Growth Factor Binding Protein 6 (IGFBP6) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54470
The Mouse Insulin Like Growth Factor Binding Protein 6 (IGFBP6) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 6.25-400 ng/mL. Sensitivity: 2.7 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P47880. NCBI Gene ID: 16012. Restriction: For Research Use Only.
Catalog No. 50-150-1345