Learn More
Biomatik Corporation Mouse Immunoglobulin G (IgG) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54475
The Mouse Immunoglobulin G (IgG) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 1.56-100 μg/mL. Sensitivity: 0.44 μg/mL. Format: 96-Well Plate. Assay type: competitive inhibition. Restriction: For Research Use Only.
Catalog No. 50-150-1348
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.