Learn More
Biomatik Corporation Human Interleukin 10 Receptor Alpha (IL10Ra) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54488
The Human Interleukin 10 Receptor Alpha (IL10Ra) ELISA Kit allows for quantitative determination in tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 31.25-2000 pg/mL. Sensitivity: 13.8 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q13651. NCBI Gene ID: 3587. Restriction: For Research Use Only.
Catalog No. 50-150-1354