Learn More
Biomatik Corporation Human Interleukin 13 (IL13) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54512
The Human Interleukin 13 (IL13) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 7.1 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P35225. NCBI Gene ID: 3596. Restriction: For Research Use Only.
Catalog No. 50-150-1368