Learn More
Biomatik Corporation Mouse Interleukin 13 Receptor Alpha 2 (IL13Ra2) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54517
The Mouse Interleukin 13 Receptor Alpha 2 (IL13Ra2) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.126 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O88786. NCBI Gene ID: 16165. Restriction: For Research Use Only.
Catalog No. 50-150-1372