Learn More
Biomatik Corporation Human Interleukin 17 Receptor A (IL17RA) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54545
The Human Interleukin 17 Receptor A (IL17RA) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 78.125-5000 pg/mL. Sensitivity: 30 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q96F46. NCBI Gene ID: 23765. Restriction: For Research Use Only.
Catalog No. 50-150-1388