Learn More
Biomatik Corporation Human Interleukin 18 Receptor 1 (IL18R1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54559
The Human Interleukin 18 Receptor 1 (IL18R1) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q13478. NCBI Gene ID: 8809. Restriction: For Research Use Only.
Catalog No. 50-150-1393