Learn More
Biomatik Corporation Rat Interleukin 19 (IL19) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54563
The Rat Interleukin 19 (IL19) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.8 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: D3ZFI7. NCBI Gene ID: 681145. Restriction: For Research Use Only.
Catalog No. 50-150-1397