Learn More
Biomatik Corporation Canine Interleukin 1 Beta (IL1b) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54575
The Canine Interleukin 1 Beta (IL1b) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine Dog. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.4 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q2VCE2. NCBI Gene ID: 403974. Restriction: For Research Use Only.
Catalog No. 50-150-1401