Learn More
Biomatik Corporation Human Interleukin 23 Receptor (IL23R) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54631
The Human Interleukin 23 Receptor (IL23R) ELISA Kit allows for quantitative determination in tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.041 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q5VWK5. NCBI Gene ID: 149233. Restriction: For Research Use Only.
Catalog No. 50-150-1429