Learn More
Biomatik Corporation Mouse Interleukin 25 (IL25) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54637
The Mouse Interleukin 25 (IL25) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.7 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q8VHH8. NCBI Gene ID: 140806. Restriction: For Research Use Only.
Catalog No. 50-150-1431