Learn More
Biomatik Corporation Human Interleukin 2 Receptor Alpha (IL2Ra) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54647
The Human Interleukin 2 Receptor Alpha (IL2Ra) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.051 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P01589. NCBI Gene ID: 3559. Restriction: For Research Use Only.
Catalog No. 50-150-1435