Learn More
Biomatik Corporation Human Interleukin 32 (IL32) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54658
The Human Interleukin 32 (IL32) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.0 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P24001. NCBI Gene ID: 9235. Restriction: For Research Use Only.
Catalog No. 50-150-1441