Learn More
Biomatik Corporation Mouse Interleukin 33 (IL33) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54660
The Mouse Interleukin 33 (IL33) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 78.125-5000 pg/mL. Sensitivity: 31 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q8BVZ5. NCBI Gene ID: 77125. Restriction: For Research Use Only.
Catalog No. 50-150-1443