Learn More
Biomatik Corporation Canine Interleukin 8 (IL8) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54713
The Canine Interleukin 8 (IL8) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine Dog. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P41324. NCBI Gene ID: 403850. Restriction: For Research Use Only.
Catalog No. 50-150-1464