Learn More
Biomatik Corporation Guinea pig Interleukin 8 (IL8) ELISA Kit, 96 tests DFS Item
Manufacturer:  Biomatik Corporation EKL54716
The Guinea pig Interleukin 8 (IL8) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Cavia Guinea pig. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.8 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P49113. NCBI Gene ID: 100379599. Restriction: For Research Use Only.
Catalog No. 50-150-1466