Learn More
Biomatik Corporation Human Interleukin 9 (IL9) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54729
The Human Interleukin 9 (IL9) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.063 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P15248. NCBI Gene ID: 3578. Restriction: For Research Use Only.
Catalog No. 50-150-1470