Learn More
Biomatik Corporation Human Interleukin 1 Receptor Associated Kinase 1 (IRAK1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54740
The Human Interleukin 1 Receptor Associated Kinase 1 (IRAK1) ELISA Kit allows for quantitative determination in tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.105 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P51617. NCBI Gene ID: 3654. Restriction: For Research Use Only.
Catalog No. 50-150-1476