Learn More
Biomatik Corporation Human Keratin 81 (KRT81) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54764
The Human Keratin 81 (KRT81) ELISA Kit allows for quantitative determination in tissue homogenates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.053 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q14533. NCBI Gene ID: 3887. Restriction: For Research Use Only.
Catalog No. 50-150-149-0