Learn More
Biomatik Corporation Rat Nerve Growth Factor (NGF) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54831
The Rat Nerve Growth Factor (NGF) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Rattus norvegicus Rat. Detection Range: 78.1-5000 pg/mL. Sensitivity: 33.2 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P25427. NCBI Gene ID: 310738. Restriction: For Research Use Only.
Catalog No. 50-150-1529