Learn More
Biomatik Corporation Human N-Terminal Pro Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54844
The Human N-Terminal Pro Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 39.062-2500 pg/mL. Sensitivity: 15 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. Restriction: For Research Use Only.
Catalog No. 50-150-1532