Learn More
Biomatik Corporation Human Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54862
The Human Platelet Derived Growth Factor Subunit A (PDGFA) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.063 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P04085. NCBI Gene ID: 5154. Restriction: For Research Use Only.
Catalog No. 50-150-1542