Learn More
Biomatik Corporation Human Platelet Derived Growth Factor AB (PDGFAB) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54866
The Human Platelet Derived Growth Factor AB (PDGFAB) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 78.125-5000 pg/mL. Sensitivity: 27 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. Restriction: For Research Use Only.
Catalog No. 50-150-1545