Learn More
Biomatik Corporation Human Squamous Cell Carcinoma Antigen 1 (SCCA1) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54924
The Human Squamous Cell Carcinoma Antigen 1 (SCCA1) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 78.125-5000 pg/mL. Sensitivity: 34 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P29508. NCBI Gene ID: 6317. Restriction: For Research Use Only.
Catalog No. 50-150-1567