Learn More
Biomatik Corporation Human Transforming Growth Factor Beta Induced Protein (TGFbI) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54985
The Human Transforming Growth Factor Beta Induced Protein (TGFbI) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.625-40 ng/mL. Sensitivity: 0.245 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: Q15582. NCBI Gene ID: 7045. Restriction: For Research Use Only.
Catalog No. 50-150-1587