Learn More
Biomatik Corporation Mouse Transforming Growth Factor Beta Induced Protein (TGFbI) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL54986
The Mouse Transforming Growth Factor Beta Induced Protein (TGFbI) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 62.5-4000 pg/mL. Sensitivity: 22.5 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P82198. NCBI Gene ID: 21810. Restriction: For Research Use Only.
Catalog No. 50-150-1588