Learn More
Biomatik Corporation Human Tumor Necrosis Factor Beta (TNFb) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL55013
The Human Tumor Necrosis Factor Beta (TNFb) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 15.625-1000 pg/mL. Sensitivity: 6.2 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P01374. NCBI Gene ID: 4049. Restriction: For Research Use Only.
Catalog No. 50-150-1601