Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Receptor Superfamily, Member 17 (TNFRSF17) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL55018
The Mouse Tumor Necrosis Factor Receptor Superfamily, Member 17 (TNFRSF17) ELISA Kit allows for quantitative determination in serum, plasma and other biological fluids. Species Reactivity: Mus musculus mouse. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.101 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: O88472. NCBI Gene ID: 21935. Restriction: For Research Use Only.
Catalog No. 50-150-1604