Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily, Member 1A (TNFRSF1A) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL55019
The Human Tumor Necrosis Factor Receptor Superfamily, Member 1A (TNFRSF1A) ELISA Kit allows for quantitative determination in serum, plasma, urine, cell lysates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 78.125-5000 pg/mL. Sensitivity: 35 pg/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P19438. NCBI Gene ID: 7132. Restriction: For Research Use Only.
Catalog No. 50-150-1605