Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily, Member 1B (TNFRSF1B) ELISA Kit, 96 tests  DFS Item
Manufacturer:  Biomatik Corporation EKL55021
The Human Tumor Necrosis Factor Receptor Superfamily, Member 1B (TNFRSF1B) ELISA Kit allows for quantitative determination in serum, plasma, tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens human. Detection Range: 0.156-10 ng/mL. Sensitivity: 0.063 ng/mL. Format: 96-Well Plate. Assay type: Sandwich. UniProt ID: P20333. NCBI Gene ID: 7133. Restriction: For Research Use Only.
Catalog No. 50-150-1606